siRNA Details
Gene Symbol: VENTX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014468.3 | NP_055283.1 | 2 | 402 | ||
XM_017016073.1 | XP_016871562.1 | 2 | 397 | |||
GenBank mRNA | AF068006.1 | - | 2 | 402 | ||
BC108915.1 | - | 2 | 402 | 40015551 | ||
BC108916.1 | - | 2 | 402 | 40015553 |
Gene Symbol: VENTXP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002311.1 | 1 | 399 | |||
GenBank mRNA | X74864.1 | - | 1 | 97 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
