siRNA Details
Gene Symbol: CFAP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164496.1 | NP_001157968.1 | 20 | 2806 | ||
NM_018338.3 | NP_060808.2 | 20 | 2806 | |||
XM_005247617.3 | XP_005247674.1 | 3 | 1083 | |||
XM_006713696.2 | XP_006713759.1 | 21 | 3396 | |||
XM_011512977.2 | XP_011511279.1 | 21 | 3396 | |||
XM_011512978.2 | XP_011511280.1 | 16 | 2293 | |||
XM_011512979.2 | XP_011511281.1 | 15 | 2037 | |||
XM_011512980.2 | XP_011511282.1 | 21 | 3396 | |||
XM_011512981.1 | XP_011511283.1 | 21 | 3396 | |||
XM_011512982.2 | XP_011511284.1 | 21 | 3396 | |||
XM_011512983.2 | XP_011511285.1 | 21 | 3396 | |||
XM_011512984.2 | XP_011511286.1 | 21 | 3396 | |||
XM_017006842.1 | XP_016862331.1 | 21 | 3396 | |||
XM_017006843.1 | XP_016862332.1 | 21 | 3396 | |||
XM_017006844.1 | XP_016862333.1 | 20 | 3240 | |||
XM_017006846.1 | XP_016862335.1 | 15 | 3191 | |||
XM_017006847.1 | XP_016862336.1 | 16 | 2234 | |||
XM_017006848.1 | XP_016862337.1 | 21 | 3396 | |||
XM_017006849.1 | XP_016862338.1 | 21 | 3396 | |||
GenBank mRNA | AK056472.1 | - | 20 | 2785 | ||
AL133606.1 | - | 2 | 148 | |||
BC101582.1 | - | 20 | 2764 | 8069088 | ||
BC113553.1 | - | 20 | 2765 | 8322605 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
