siRNA Details
Gene Symbol: FAM86DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024241.1 | 3 | 301 | |||
GenBank mRNA | BC043342.2 | - | 3 | 127 | ||
BC068108.1 | - | 3 | 234 | |||
BC094682.1 | - | 3 | 348 | |||
DA753838.1 | - | 3 | 301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
