siRNA Details
Gene Symbol: SLC35G3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152462.2 | NP_689675.1 | 1 | 724 | ||
GenBank mRNA | AK097473.1 | - | 1 | 689 | ||
BC136422.1 | - | 1 | 724 | 9020411 | ||
BC140700.1 | - | 1 | 724 | 9020410 |
Gene Symbol: SLC35G4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282300.1 | NP_001269229.1 | 1 | 676 |
Gene Symbol: SLC35G5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054028.1 | NP_473369.1 | 1 | 759 | ||
GenBank mRNA | AJ291677.1 | - | 1 | 759 | ||
BC131696.1 | - | 1 | 726 | 40120574 |
Gene Symbol: SLC35G6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102614.1 | NP_001096084.1 | 2 | 753 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
