siRNA Details
Gene Symbol: DNAAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000404.1 | - | 4, 5 | 753 | ||
AK054595.1 | - | 4, 5 | 1184 | |||
AK096700.1 | - | 10, 11 | 1603 | |||
AK225507.1 | - | 4, 5 | 753 | |||
AL832914.1 | - | 2, 3 | 253 | |||
BC010850.1 | - | 6, 7 | 983 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
