siRNA Details
Gene Symbol: KGFLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003674.2 | 2 | 646 | |||
NR_047527.1 | 3 | 756 | ||||
GenBank mRNA | AF523265.1 | - | 2 | 741 | ||
AK054997.1 | - | 2 | 385 | |||
AK307215.1 | - | 2 | 377 | |||
AY098593.1 | - | 2 | 637 | |||
BC112977.1 | - | 2 | 644 | |||
BC112978.1 | - | 2 | 644 | |||
DB501751.1 | - | 3 | 297 |
Gene Symbol: KGFLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003670.1 | 2 | 745 | |||
GenBank mRNA | AY098594.1 | - | 2 | 637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
