siRNA Details
Gene Symbol: CDC26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139286.3 | NP_644815.1 | 3 | 401 | ||
XM_017014573.1 | XP_016870062.1 | 3 | 368 | |||
XM_017014574.1 | XP_016870063.1 | 3 | 833 | |||
XM_017014575.1 | XP_016870064.1 | 3 | 363 | |||
GenBank mRNA | AF503918.1 | - | 1 | 42 | ||
BC042534.1 | - | 3 | 356 | 4751173 | ||
BC066300.1 | - | 3 | 386 | 5258904 | ||
BF244254.1 | - | 3 | 398 | |||
CD685602.1 | - | 3 | 406 |
Gene Symbol: FKBP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK307507.1 | - | 3 | 253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
