siRNA Details
Gene Symbol: CDC26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139286.3 | NP_644815.1 | 4 | 599 | ||
XM_017014573.1 | XP_016870062.1 | 4 | 566 | |||
XM_017014574.1 | XP_016870063.1 | 4 | 1031 | |||
XM_017014575.1 | XP_016870064.1 | 4 | 561 | |||
GenBank mRNA | AF503918.1 | - | 1 | 240 | ||
BC042534.1 | - | 4 | 554 | 4751173 | ||
BC066300.1 | - | 4 | 584 | 5258904 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
