siRNA Details
Gene Symbol: NCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF330625.1 | - | 3 | 254 | ||
AF330626.1 | - | 3 | 190 |
Gene Symbol: NCF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003186.1 | 4 | 323 | |||
GenBank mRNA | AK298655.1 | - | 6 | 581 | ||
BC080194.1 | - | 4 | 323 |
Gene Symbol: NCF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003187.3 | 6 | 613 | |||
GenBank mRNA | AF330627.1 | - | 6 | 570 | ||
AK298739.1 | - | 5 | 503 | |||
AK301397.1 | - | 4 | 338 | |||
AK316329.1 | - | 6 | 613 | |||
M25665.1 | - | 6 | 572 | |||
M55067.1 | - | 6 | 582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
