siRNA Details
Gene Symbol: BRD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173984.2 | NP_001167455.1 | 2, 3 | 419 | ||
NM_013263.4 | NP_037395.2 | 2, 3 | 419 | |||
XM_017023179.1 | XP_016878668.1 | 2, 3 | 319 | |||
XM_017023180.1 | XP_016878669.1 | 2, 3 | 319 | |||
GenBank mRNA | AF152604.1 | - | 2, 3 | 312 | ||
AF213969.1 | - | 2, 3 | 264 | |||
AJ271881.1 | - | 2, 3 | 377 | |||
BC050728.1 | - | 3, 4 | 326 | 6149862 | ||
BC094706.1 | - | 3, 4 | 296 | 30719813 | ||
DA645362.1 | - | 2, 3 | 319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
