siRNA Details
Gene Symbol: C4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252204.1 | NP_001239133.1 | 28 | 3580 | ||
NM_007293.2 | NP_009224.2 | 28 | 3580 | |||
GenBank mRNA | BC063289.1 | - | 28 | 3613 | 5289068 | |
BC144546.1 | - | 28 | 3550 | 9053073 | ||
BC146673.1 | - | 28 | 3569 | 40148477 | ||
BC146849.1 | - | 28 | 3550 | 9056777 | ||
BC151204.1 | - | 28 | 3567 | 40147083 | ||
BC171786.1 | - | 28 | 3550 | 9054440 | ||
K02403.1 | - | 28 | 3569 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
