siRNA Details
Gene Symbol: RTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283035.1 | NP_001269964.1 | 10 | 1547 | ||
NM_001283036.1 | NP_001269965.1 | 9 | 1454 | |||
NM_001283037.1 | NP_001269966.1 | 8 | 1402 | |||
NM_016407.4 | NP_057491.2 | 9 | 1457 | |||
XM_017027872.1 | XP_016883361.1 | 12 | 1650 | |||
GenBank mRNA | AF117231.1 | - | 9 | 1407 | ||
AF161513.1 | - | 9 | 1417 | |||
AF161518.1 | - | 9 | 1383 | |||
AF212244.1 | - | 9 | 1395 | |||
AK000481.1 | - | 9 | 1387 | |||
AK000586.1 | - | 8 | 1350 | |||
BC003359.1 | - | 9 | 1384 | 2900332 |
