siRNA Details
Gene Symbol: GOLGA6D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145224.1 | NP_001138696.1 | 6 | 426 |
Gene Symbol: GOLGA6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038640.2 | NP_001033729.2 | 6 | 432 | ||
GenBank mRNA | AF263742.1 | - | 6 | 432 | ||
BC131515.1 | - | 6 | 412 | |||
DB039513.1 | - | 6 | 431 |
Gene Symbol: GOLGA6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018652.4 | NP_061122.4 | 6 | 432 | ||
XM_017022406.1 | XP_016877895.1 | 11 | 1320 |
Gene Symbol: GOLGA6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164404.1 | NP_001157876.1 | 6 | 432 | ||
XM_017022519.1 | XP_016878008.1 | 6 | 586 |
Gene Symbol: LOC645752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027024.1 | 6 | 417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
