siRNA Details
Gene Symbol: ARHGAP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199417.1 | NP_001186346.1 | 15 | 2687 | ||
XM_006721991.1 | XP_006722054.1 | 15 | 2728 | |||
XM_006721992.3 | XP_006722055.1 | 15 | 2466 | |||
XM_006721993.3 | XP_006722056.1 | 15 | 2714 | |||
XM_006721994.3 | XP_006722057.1 | 15 | 2713 | |||
XM_011525073.1 | XP_011523375.1 | 15 | 2647 | |||
XM_011525074.2 | XP_011523376.1 | 15 | 2746 | |||
XM_011525075.2 | XP_011523377.1 | 10 | 2288 | |||
XM_011525076.2 | XP_011523378.1 | 15 | 2713 | |||
XM_011525077.2 | XP_011523379.1 | 15 | 2713 | |||
XM_017024902.1 | XP_016880391.1 | 15 | 2713 | |||
GenBank mRNA | AB040934.1 | - | 9 | 1450 | ||
BC039190.1 | - | 8 | 960 | |||
DN831435.1 | - | 2 | 163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
