siRNA Details
Gene Symbol: ARHGAP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199417.1 | NP_001186346.1 | 11 | 2051 | ||
XM_006721991.1 | XP_006722054.1 | 11 | 2092 | |||
XM_006721992.3 | XP_006722055.1 | 11 | 2079 | |||
XM_006721993.3 | XP_006722056.1 | 11 | 2078 | |||
XM_006721994.3 | XP_006722057.1 | 11 | 2077 | |||
XM_011525073.1 | XP_011523375.1 | 11 | 2011 | |||
XM_011525074.2 | XP_011523376.1 | 11 | 2110 | |||
XM_011525075.2 | XP_011523377.1 | 6 | 1652 | |||
XM_011525076.2 | XP_011523378.1 | 11 | 2077 | |||
XM_011525077.2 | XP_011523379.1 | 11 | 2077 | |||
XM_017024902.1 | XP_016880391.1 | 11 | 2077 | |||
GenBank mRNA | AB040934.1 | - | 5 | 814 | ||
BC039190.1 | - | 4 | 324 | |||
BM548592.1 | - | 5 | 585 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_021493, CB525670, BQ444174.
Gene Ontology Categories:
