siRNA Details
Gene Symbol: LINC00671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027254.1 | 2 | 117 | |||
GenBank mRNA | AK055784.1 | - | 3 | 319 | ||
BC122867.1 | - | 2 | 39 | |||
BC122868.1 | - | 2 | 39 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
