siRNA Details
Gene Symbol: LINC00671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027254.1 | 4 | 444 | |||
GenBank mRNA | AK055784.1 | - | 4 | 554 | ||
AK298293.1 | - | 3 | 375 | |||
BC122867.1 | - | 3 | 274 | |||
BC122868.1 | - | 4 | 366 | |||
DC361857.1 | - | 3 | 375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
