siRNA Details
Gene Symbol: FTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002032.2 | NP_002023.2 | 2 | 381 | ||
GenBank mRNA | AB062402.1 | - | 2 | 334 | ||
AF088851.1 | - | 2 | 188 | |||
AK311818.1 | - | 2 | 352 | |||
AY258285.1 | - | 939 | ||||
BC000857.1 | - | 2 | 340 | 3459353 | ||
BC001399.2 | - | 2 | 309 | 3051458 | ||
BC011359.2 | - | 2 | 331 | 4157296 | ||
BC013724.2 | - | 2 | 334 | 3857790 | ||
BC015156.1 | - | 2 | 204 | 3883694 | ||
BC016009.1 | - | 2 | 356 | 4666545 | ||
BC016857.1 | - | 2 | 300 | 3848510 | ||
BC020300.1 | - | 2 | 317 | |||
BC035441.1 | - | 2 | 340 | |||
BC063514.1 | - | 2 | 323 | 3882345 | ||
BC066341.1 | - | 3 | 258 | 5288688 | ||
BC066961.1 | - | 2 | 354 | 4796312 | ||
BC070494.1 | - | 3 | 295 | |||
BC073750.1 | - | 2 | 338 | 6256153 | ||
BC104643.1 | - | 2 | 356 | 4043391 | ||
BC105802.1 | - | 3 | 333 | |||
BQ887222.1 | - | 381 | ||||
L20941.1 | - | 2 | 354 | |||
M11146.1 | - | 2 | 223 | |||
M12937.1 | - | 2 | 172 | |||
M97164.1 | - | 2 | 275 | |||
X00318.1 | - | 2 | 237 |
Gene Symbol: FTH1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002201.1 | 1 | 355 | |||
GenBank mRNA | BC073777.1 | - | 1 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
