siRNA Details
Gene Symbol: RPSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304288.1 | NP_001291217.1 | 7 | 1069 | ||
NM_002295.5 | NP_002286.2 | 7 | 1054 | |||
GenBank mRNA | AF503367.1 | - | 1 | 1014 | ||
AJ572067.1 | - | 1 | 131 | |||
BC005391.1 | - | 1 | 1026 | 3997019 | ||
BC008867.2 | - | 1 | 975 | 4130936 | ||
BC010418.2 | - | 1 | 1006 | 4079845 | ||
BC013827.2 | - | 1 | 1010 | 3446816 | ||
BC018867.2 | - | 1 | 1000 | |||
BC034537.1 | - | 1 | 1007 | 3455545 | ||
BC050688.1 | - | 1 | 1009 | 4443949 | ||
BC053370.1 | - | 1 | 1007 | 5108337 | ||
BC066941.1 | - | 7 | 1023 | 4820432 | ||
BC068062.1 | - | 1 | 988 | 5427114 | ||
BC071693.1 | - | 1 | 1016 | 3884457 | ||
BC071968.1 | - | 1 | 1007 | 5814758 | ||
BC071969.1 | - | 1 | 995 | 6483856 | ||
BC071970.1 | - | 1 | 980 | 6484091 | ||
BC073863.1 | - | 1 | 992 | 3924066 | ||
M14199.1 | - | 4 | 465 |
Gene Symbol: FAM149A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008008.1 | XP_016863497.1 | 9 | 2132 |
Gene Symbol: RPSAP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648992.1 | - | 2 | 2500 |
Gene Symbol: RPSAP52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026825.2 | 2 | 1103 | |||
GenBank mRNA | BC058822.1 | - | 2 | 1107 | ||
BC107865.1 | - | 2 | 1107 |
Gene Symbol: RPSAP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003662.3 | 4 | 1368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
