siRNA Details
Gene Symbol: SPDYE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031618.3 | NP_001026789.2 | 7 | 1312 | ||
XM_011516237.2 | XP_011514539.1 | 7 | 1311 | |||
XM_017012223.1 | XP_016867712.1 | 7 | 1311 | |||
GenBank mRNA | AK302217.1 | - | 7 | 1312 | ||
BC100973.1 | - | 4 | 550 | 40005226 | ||
BF809327.1 | - | 1 | 232 | |||
DR731150.1 | - | 4 | 456 | |||
JQ023160.1 | - | 6 | 838 |
Gene Symbol: SPDYE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175064.2 | NP_778234.2 | 5 | 851 | ||
XM_005249719.3 | XP_005249776.1 | 7 | 1362 | |||
XM_017012015.1 | XP_016867504.1 | 7 | 1300 | |||
XM_017012016.1 | XP_016867505.1 | 6 | 712 | |||
XM_017012017.1 | XP_016867506.1 | 4 | 947 | |||
GenBank mRNA | AF412027.1 | - | 5 | 860 | ||
AL137266.1 | - | 5 | 633 |
Gene Symbol: SPDYE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166339.1 | NP_001159811.1 | 7 | 1312 | ||
XM_005250093.3 | XP_005250150.1 | 6 | 1880 | |||
XM_011515702.2 | XP_011514004.1 | 6 | 1880 |
Gene Symbol: SPDYE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306141.1 | NP_001293070.1 | 6 | 1259 |
Gene Symbol: SPDYE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146210.2 | NP_001139682.2 | 6 | 1259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
