siRNA Details
Gene Symbol: UBA52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033930.2 | NP_001029102.1 | 3, 4 | 298 | ||
NM_001321017.1 | NP_001307946.1 | 3, 4 | 297 | |||
NM_001321018.1 | NP_001307947.1 | 3, 4 | 924 | |||
NM_001321019.1 | NP_001307948.1 | 3, 4 | 462 | |||
NM_001321020.1 | NP_001307949.1 | 3, 4 | 496 | |||
NM_001321021.1 | NP_001307950.1 | 3, 4 | 311 | |||
NM_003333.4 | NP_003324.1 | 3, 4 | 276 | |||
XM_005260052.2 | XP_005260109.1 | 3, 4 | 242 | |||
XM_005260053.3 | XP_005260110.1 | 3, 4 | 274 | |||
XM_005260054.2 | XP_005260111.1 | 2, 3 | 204 | |||
XM_006722871.2 | XP_006722934.1 | 3, 4 | 928 | |||
XM_017027198.1 | XP_016882687.1 | 3, 4 | 1251 | |||
GenBank mRNA | AF348700.1 | - | 3, 4 | 223 | ||
AK314717.1 | - | 3, 4 | 208 | |||
AV707734.1 | - | 276 | ||||
BC101830.1 | - | 3, 4 | 240 | 8069336 | ||
BC101832.1 | - | 3, 4 | 240 | 8069338 | ||
BC143668.1 | - | 3, 4 | 240 | 9052182 | ||
BE514609.1 | - | 3, 4 | 443 | |||
BG341101.1 | - | 3, 4 | 314 | |||
BQ428515.1 | - | 2, 3 | 205 | |||
BQ428688.1 | - | 278 | ||||
CB124676.1 | - | 3, 4 | 242 | |||
CD242660.1 | - | 3, 4 | 503 | |||
CD385706.1 | - | 3, 4 | 467 | |||
DA949569.1 | - | 3, 4 | 297 | |||
HY153317.1 | - | 3, 4 | 275 | |||
X56998.1 | - | 2, 3 | 191 | |||
X56999.1 | - | 2, 3 | 200 | |||
Y00361.1 | - | 1, 2 | 66 |
Gene Symbol: UBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281716.1 | NP_001268645.1 | 2 | 888 | ||
NM_001281717.1 | NP_001268646.1 | 2 | 859 | |||
NM_001281718.1 | NP_001268647.1 | 2 | 819 | |||
NM_001281719.1 | NP_001268648.1 | 2 | 817 | |||
NM_001281720.1 | NP_001268649.1 | 2 | 663 | |||
NM_018955.3 | NP_061828.1 | 2 | 1032 | |||
GenBank mRNA | AK129498.1 | - | 2 | 734 | ||
AK300882.1 | - | 3 | 510 | |||
BC000379.2 | - | 2 | 776 | 2820408 | ||
BC009301.2 | - | 2 | 722 | 4126622 | ||
BC015127.1 | - | 2 | 738 | 3929901 | ||
BC026301.1 | - | 2 | 734 | 4280629 | ||
BC031027.1 | - | 2 | 736 | 4715377 | ||
BC038999.1 | - | 2 | 745 | 4943285 | ||
BC046123.2 | - | 2 | 724 | 3879581 | ||
BM554254.1 | - | 2 | 742 | |||
BT020104.1 | - | 1 | 638 | |||
GQ472200.1 | - | 2 | 776 |
Gene Symbol: UBC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021009.6 | NP_066289.3 | 2 | 640 | ||
GenBank mRNA | AB009010.1 | - | 2 | 250 | ||
AB209782.1 | - | 230 | ||||
AF052164.1 | - | 1 | 107 | |||
AK026846.1 | - | 2 | 261 | |||
AK056939.1 | - | 2, 3 | 249 | |||
AK092983.1 | - | 1754 | ||||
AK097415.1 | - | 1 | 378 | |||
AK098827.1 | - | 2 | 249 | |||
AK129749.1 | - | 2 | 249 | |||
AK291539.1 | - | 2 | 249 | |||
AL117514.1 | - | 1 | 212 | |||
AY732488.1 | - | 1 | 200 | |||
AY819759.1 | - | 1 | 200 | |||
BC000449.2 | - | 2 | 235 | |||
BC008955.2 | - | 2 | 223 | |||
BC014880.1 | - | 1 | 73 | |||
BC039193.1 | - | 2 | 250 | 4076286 | ||
BC080583.1 | - | 2 | 223 | |||
BC093445.1 | - | 2 | 249 | |||
BE873233.1 | - | 460 | ||||
BF793240.1 | - | 1 | 125 | |||
BU195276.1 | - | 2 | 235 | |||
M17597.1 | - | 1 | 74 | |||
M26880.1 | - | 317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
