siRNA Details
Gene Symbol: DEFB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205266.1 | NP_001192195.1 | 2 | 260 | ||
GenBank mRNA | BE714502.1 | - | 2 | 248 |
Gene Symbol: DEFB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004942.3 | NP_004933.1 | 2 | 403 | ||
GenBank mRNA | Z71389.1 | - | 2 | 278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
