siRNA Details
Gene Symbol: OR5T3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004747.1 | NP_001004747.1 | 1 | 322 |
Gene Symbol: OR5T1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004745.1 | NP_001004745.1 | 1 | 268 | ||
GenBank mRNA | BC136992.1 | - | 1 | 354 | 9020984 | |
BC136993.1 | - | 1 | 354 | 9020985 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
