siRNA Details
Gene Symbol: SNRPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003091.3 | NP_003082.1 | 2 | 248 | ||
NM_198216.1 | NP_937859.1 | 2 | 248 | |||
GenBank mRNA | AK301034.1 | - | 2 | 180 | ||
AK301206.1 | - | 2 | 182 | |||
AK307093.1 | - | 2 | 182 | |||
AK307266.1 | - | 2 | 182 | |||
AK312620.1 | - | 2 | 183 | |||
AY742712.1 | - | 1 | 85 | |||
BC080516.1 | - | 2 | 166 | 2822685 | ||
BM559678.1 | - | 248 | ||||
CR456969.1 | - | 1 | 85 | |||
J04564.1 | - | 2 | 211 | |||
M34081.1 | - | 1 | 64 | |||
M34082.1 | - | 1 | 64 | |||
X15893.1 | - | 1 | 85 | |||
X17567.1 | - | 2 | 139 | |||
X17568.1 | - | 2 | 124 |

More Information
Other Species Targeted

Gene Ontology Categories:
