siRNA Details
Gene Symbol: SNRPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003091.3 | NP_003082.1 | 2 | 303 | ||
NM_198216.1 | NP_937859.1 | 2 | 303 | |||
GenBank mRNA | AK301034.1 | - | 2 | 235 | ||
AK301206.1 | - | 2 | 237 | |||
AK307093.1 | - | 2 | 237 | |||
AK307266.1 | - | 2 | 237 | |||
AK312620.1 | - | 2 | 238 | |||
AY742712.1 | - | 1 | 140 | |||
BC080516.1 | - | 2 | 221 | 2822685 | ||
BM559678.1 | - | 303 | ||||
CR456969.1 | - | 1 | 140 | |||
J04564.1 | - | 2 | 266 | |||
M34081.1 | - | 1 | 119 | |||
M34082.1 | - | 1 | 119 | |||
X15893.1 | - | 1 | 140 | |||
X17567.1 | - | 2 | 194 | |||
X17568.1 | - | 2 | 179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
