siRNA Details
Gene Symbol: THOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032361.3 | NP_115737.1 | 2 | 443 | ||
XM_017009985.1 | XP_016865474.1 | 3 | 310 | |||
GenBank mRNA | BC006849.1 | - | 2 | 405 | 3451612 | |
BC066325.1 | - | 2 | 400 | 5303134 | ||
BC068499.1 | - | 2 | 362 | 5262880 | ||
BG502664.1 | - | 2 | 164 | |||
BU781626.1 | - | 1 | 40 | |||
HY009859.1 | - | 2 | 444 |
Gene Symbol: LOC728554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003615.2 | 2 | 669 | |||
GenBank mRNA | AK310274.1 | - | 2 | 143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
