siRNA Details
Gene Symbol: RPL10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007104.4 | NP_009035.3 | 5 | 505 | ||
XM_017010900.1 | XP_016866389.1 | 5 | 760 | |||
GenBank mRNA | AB082926.1 | - | 1 | 486 | ||
AF086557.1 | - | 1 | 134 | |||
AK092386.1 | - | 1 | 930 | |||
AK313181.1 | - | 5 | 504 | |||
BC006791.1 | - | 5 | 486 | 2901249 | ||
BC011366.1 | - | 4 | 477 | 2986088 | ||
BC070216.1 | - | 1 | 493 | 6495090 | ||
BC116456.1 | - | 1 | 438 | |||
BT020040.1 | - | 4 | 471 | |||
BT020041.1 | - | 4 | 471 | |||
U12404.1 | - | 5 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
