siRNA Details
Gene Symbol: DAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004081.5 | NP_004072.3 | 3, 4 | 539 | ||
XM_011531482.2 | XP_011529784.1 | 3, 4 | 577 | |||
XM_011531483.1 | XP_011529785.1 | 2, 3 | 199 | |||
GenBank mRNA | AF271087.1 | - | 3, 4 | 381 | ||
AF271088.1 | - | 14 | ||||
BC114927.1 | - | 3, 4 | 370 | 40028543 |
Gene Symbol: DAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005785.2 | NP_001005785.1 | 3, 4 | 531 | ||
NM_001005786.2 | NP_001005786.2 | 3, 4 | 531 | |||
NM_020363.3 | NP_065096.2 | 3, 4 | 531 | |||
GenBank mRNA | AF248480.1 | - | 3, 4 | 443 | ||
AF414184.1 | - | 3, 4 | 513 | |||
AK302398.1 | - | 3, 4 | 454 | |||
AK302690.1 | - | 2, 3 | 384 | |||
BC113006.1 | - | 3, 4 | 339 | 40035862 | ||
U21663.1 | - | 3, 4 | 455 |
Gene Symbol: DAZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020364.3 | NP_065097.2 | 3, 4 | 554 | ||
GenBank mRNA | AF248481.1 | - | 3, 4 | 435 | ||
BC113005.1 | - | 3, 4 | 339 | 40035860 |
Gene Symbol: DAZ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005375.2 | NP_001005375.1 | 3, 4 | 554 | ||
NM_020420.3 | NP_065153.1 | 3, 4 | 554 | |||
XM_005262602.1 | XP_005262659.1 | 3, 4 | 493 | |||
XM_011531509.2 | XP_011529811.1 | 3, 4 | 580 | |||
XM_011531510.1 | XP_011529812.1 | 2, 3 | 199 | |||
GenBank mRNA | AF248482.1 | - | 3, 4 | 443 | ||
AF248483.1 | - | 2, 3 | 244 | |||
BC047480.1 | - | 3, 4 | 459 | 5297459 | ||
BC047617.1 | - | 3, 4 | 455 | 4836748 |
Gene Symbol: DAZL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190811.1 | NP_001177740.1 | 3, 4 | 595 | ||
NM_001351.3 | NP_001342.2 | 3, 4 | 541 | |||
GenBank mRNA | AK303026.1 | - | 3, 4 | 595 | ||
AK310341.1 | - | 3, 4 | 316 | |||
BC027595.2 | - | 3, 4 | 541 | 4829381 | ||
BX648638.1 | - | 3, 4 | 543 | |||
U65918.1 | - | 3, 4 | 463 | |||
U66078.1 | - | 3, 4 | 473 | |||
U66726.2 | - | 3, 4 | 463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
