siRNA Details
Gene Symbol: DAZ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005375.2 | NP_001005375.1 | 5 | 627 | ||
NM_020420.3 | NP_065153.1 | 5 | 627 | |||
XM_005262602.1 | XP_005262659.1 | 5 | 566 | |||
XM_011531509.2 | XP_011529811.1 | 5 | 653 | |||
XM_011531510.1 | XP_011529812.1 | 4 | 272 | |||
GenBank mRNA | AF248482.1 | - | 5 | 516 | ||
AF248483.1 | - | 4 | 317 | |||
BC047480.1 | - | 5 | 532 | 5297459 | ||
BC047617.1 | - | 5 | 528 | 4836748 |
Gene Symbol: DAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004081.5 | NP_004072.3 | 5 | 612 | ||
XM_011531482.2 | XP_011529784.1 | 5 | 650 | |||
XM_011531483.1 | XP_011529785.1 | 4 | 272 | |||
GenBank mRNA | AF271087.1 | - | 4 | 890 | ||
AF271088.1 | - | 2 | 87 | |||
BC114927.1 | - | 5 | 443 | 40028543 |
Gene Symbol: DAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005785.2 | NP_001005785.1 | 5 | 604 | ||
NM_001005786.2 | NP_001005786.2 | 5 | 604 | |||
NM_020363.3 | NP_065096.2 | 5 | 604 | |||
GenBank mRNA | AF248480.1 | - | 5 | 516 | ||
AF414184.1 | - | 5 | 586 | |||
AK302398.1 | - | 5 | 527 | |||
AK302690.1 | - | 4 | 457 | |||
BC113006.1 | - | 5 | 412 | 40035862 | ||
U21663.1 | - | 5 | 528 |
Gene Symbol: DAZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020364.3 | NP_065097.2 | 5 | 627 | ||
GenBank mRNA | AF248481.1 | - | 5 | 508 | ||
BC113005.1 | - | 5 | 412 | 40035860 |
Gene Symbol: DAZL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190811.1 | NP_001177740.1 | 5 | 668 | ||
NM_001351.3 | NP_001342.2 | 5 | 614 | |||
GenBank mRNA | AK303026.1 | - | 5 | 668 | ||
AK310341.1 | - | 5 | 389 | |||
BC027595.2 | - | 5 | 614 | 4829381 | ||
BX648638.1 | - | 5 | 616 | |||
U65918.1 | - | 5 | 536 | |||
U66078.1 | - | 5 | 546 | |||
U66726.2 | - | 5 | 536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
