siRNA Details
Gene Symbol: RBMY1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006120.3 | NP_001006120.2 | 12 | 1652 | ||
NM_001320949.1 | NP_001307878.1 | 11 | 1541 | |||
NM_001320950.1 | NP_001307879.1 | 12 | 1592 | |||
XM_011531486.2 | XP_011529788.1 | 12 | 1525 | |||
XM_011531489.1 | XP_011529791.1 | 9 | 1279 | |||
XM_011531490.1 | XP_011529792.1 | 11 | 1451 | |||
GenBank mRNA | BC144422.1 | - | 11 | 1454 | 9052943 | |
BC144423.1 | - | 12 | 1505 |
Gene Symbol: RBMY1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320944.1 | NP_001307873.1 | 11 | 1541 | ||
NM_001320945.1 | NP_001307874.1 | 11 | 1474 | |||
NM_005058.3 | NP_005049.1 | 12 | 1652 | |||
XM_011531438.1 | XP_011529740.1 | 9 | 1258 | |||
XM_017030060.1 | XP_016885549.1 | 10 | 1317 | |||
GenBank mRNA | BC047768.1 | - | 11 | 1449 | 5744153 | |
BC070298.1 | - | 11 | 1506 | 6615329 | ||
BC146853.1 | - | 12 | 1565 | 9056781 | ||
BX119894.1 | - | 1 | 136 | |||
X76059.1 | - | 12 | 1617 |
Gene Symbol: RBMY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC140758.1 | - | 11 | 1454 | 9021626 |
Gene Symbol: RBMY1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303410.1 | NP_001290339.1 | 12 | 1607 | ||
NM_152585.2 | NP_689798.1 | 12 | 1612 | |||
XM_017030033.1 | XP_016885522.1 | 10 | 1317 | |||
GenBank mRNA | BC030018.2 | - | 12 | 1612 | 5269012 | |
X76060.1 | - | 12 | 1589 |
Gene Symbol: RBMY1J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006117.3 | NP_001006117.2 | 12 | 1677 | ||
XM_011531500.1 | XP_011529802.1 | 11 | 1437 | |||
XM_011531502.1 | XP_011529804.1 | 9 | 1215 | |||
XM_011531503.1 | XP_011529805.1 | 9 | 1201 | |||
XM_017030059.1 | XP_016885548.1 | 10 | 1317 | |||
GenBank mRNA | BC144577.1 | - | 12 | 1565 | 9053106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
