siRNA Details
Gene Symbol: PRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004676.2 | NP_004667.2 | 3 | 263 | ||
XM_017030088.1 | XP_016885577.1 | 2 | 2748 | |||
GenBank mRNA | AF000988.2 | - | 3 | 263 | ||
AF517635.1 | - | 3 | 263 | |||
BC113548.1 | - | 1 | 72 | 8322600 | ||
BC113550.1 | - | 1 | 72 | 8322602 | ||
BC143893.1 | - | 1 | 72 | 9052410 | ||
BC171757.1 | - | 1 | 72 | 9053258 |
Gene Symbol: PRY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002758.1 | NP_001002758.1 | 3 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
