siRNA Details
Gene Symbol: IGKV3-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF035036.1 | - | 1 | 111 | ||
AJ006111.1 | - | 501 | ||||
AJ006112.1 | - | 501 | ||||
AJ006114.1 | - | 501 | ||||
AJ006115.1 | - | 501 | ||||
AJ006117.1 | - | 501 | ||||
AJ006118.1 | - | 501 | ||||
AJ006119.1 | - | 501 | ||||
AJ006120.1 | - | 501 |
Gene Symbol: IGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC032451.1 | - | 2 | 184 | 5178085 |
Gene Symbol: IGKV3D-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF035031.1 | - | 1 | 108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
