siRNA Details
Gene Symbol: RPL18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000980.3 | NP_000971.1 | 4 | 453 | ||
GenBank mRNA | AK222647.1 | - | 4 | 409 | ||
AK297323.1 | - | 4 | 410 | |||
AK297358.1 | - | 4 | 321 | |||
AK300809.1 | - | 2 | 354 | |||
AK311774.1 | - | 4 | 409 | |||
BC007512.2 | - | 1 | 393 | 2961519 | ||
BC066319.1 | - | 1 | 413 | 5286436 | ||
BC071920.1 | - | 1 | 393 | 6293838 | ||
BI753938.1 | - | 396 | ||||
DQ593295.1 | - | 1 | 21 | |||
HY050156.1 | - | 4 | 454 | |||
L05093.1 | - | 4 | 393 | |||
X80822.1 | - | 1 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
