siRNA Details
Gene Symbol: SMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017411.3 | NP_059107.1 | 4 | 604 | ||
NM_022875.2 | NP_075013.1 | 4 | 604 | |||
NM_022876.2 | NP_075014.1 | 4 | 604 | |||
NM_022877.2 | NP_075015.1 | 4 | 604 | |||
XM_011543599.1 | XP_011541901.1 | 4 | 483 | |||
XM_017009787.1 | XP_016865276.1 | 4 | 483 | |||
GenBank mRNA | AK289669.1 | - | 4 | 487 | ||
BC000908.2 | - | 4 | 473 | 2985046 | ||
BC015308.1 | - | 4 | 471 | 4427000 | ||
BC070242.1 | - | 4 | 450 | 4673318 | ||
CN272420.1 | - | 2 | 307 | |||
JQ690864.1 | - | 2 | 193 | |||
JQ690865.1 | - | 2 | 193 | |||
JQ732167.1 | - | 2 | 193 | |||
U21914.1 | - | 4 | 438 |
Gene Symbol: SMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000344.3 | NP_000335.1 | 4 | 604 | ||
NM_001297715.1 | NP_001284644.1 | 4 | 604 | |||
NM_022874.2 | NP_075012.1 | 4 | 604 | |||
XM_011543596.1 | XP_011541898.1 | 4 | 483 | |||
XM_017009786.1 | XP_016865275.1 | 4 | 483 | |||
GenBank mRNA | BC062723.1 | - | 4 | 460 | 4250429 | |
JQ657798.1 | - | 2 | 193 | |||
JQ657799.1 | - | 2 | 193 | |||
JQ657801.1 | - | 2 | 193 | |||
JQ657802.1 | - | 2 | 193 | |||
JQ732166.1 | - | 2 | 193 | |||
U18423.1 | - | 4 | 474 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
