siRNA Details
Gene Symbol: BOLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031827.2 | NP_001026997.1 | 3 | 923 | ||
NM_001320579.1 | NP_001307508.1 | 3 | 885 | |||
NR_135304.1 | 2 | 241 | ||||
GenBank mRNA | AF060511.2 | - | 3 | 312 | ||
AK307153.1 | - | 2 | 598 | |||
BC022832.1 | - | 3 | 923 | 4647797 | ||
BC144688.1 | - | 3 | 610 | 9053222 | ||
BQ639539.1 | - | 2 | 218 | |||
BU590412.1 | - | 3 | 180 |
Gene Symbol: BOLA2-SMG1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK027255.1 | - | 2 | 599 |
Gene Symbol: BOLA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039182.2 | NP_001034271.1 | 3 | 923 | ||
NM_001320607.1 | NP_001307536.1 | 3 | 885 | |||
GenBank mRNA | BC137532.1 | - | 3 | 610 | 9021532 | |
BC137534.1 | - | 3 | 610 | 9021534 | ||
BU960186.1 | - | 3 | 248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
