siRNA Details
Gene Symbol: BOLA2-SMG1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320622.1 | NP_001307551.1 | 1 | 758 | ||
NM_001320623.1 | NP_001307552.1 | 1 | 758 | |||
NM_001320624.1 | NP_001307553.1 | 1 | 758 | |||
NM_001320625.1 | NP_001307554.1 | 1 | 758 | |||
NM_001320627.1 | NP_001307556.1 | 1 | 758 | |||
NR_135316.1 | 1 | 758 | ||||
GenBank mRNA | AF271775.1 | - | 1 | 353 | ||
AK027255.1 | - | 1 | 151 | |||
AK295153.1 | - | 1 | 62 | |||
BC062756.1 | - | 1 | 139 | |||
BC130401.1 | - | 1 | 445 | 40146432 | ||
BC130403.1 | - | 1 | 445 | 40146434 | ||
BG030350.1 | - | 109 | ||||
BI517472.1 | - | 1 | 108 | |||
BM451872.1 | - | 1 | 136 | |||
DA438012.1 | - | 1 | 150 |
Gene Symbol: BOLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031827.2 | NP_001026997.1 | 1 | 758 | ||
NM_001320579.1 | NP_001307508.1 | 1 | 758 | |||
GenBank mRNA | AF060511.2 | - | 1 | 147 | ||
AK307153.1 | - | 1 | 150 | |||
BC022832.1 | - | 1 | 758 | 4647797 | ||
BC144688.1 | - | 1 | 445 | 9053222 | ||
BU590412.1 | - | 1 | 15 |
Gene Symbol: BOLA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039182.2 | NP_001034271.1 | 1 | 758 | ||
NM_001320607.1 | NP_001307536.1 | 1 | 758 | |||
GenBank mRNA | BC137532.1 | - | 1 | 445 | 9021532 | |
BC137534.1 | - | 1 | 445 | 9021534 | ||
BU960186.1 | - | 1 | 121 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
