siRNA Details
Gene Symbol: TRMT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020810.3 | NP_065861.3 | 5 | 4733 | ||
XM_005267916.3 | XP_005267973.2 | 5 | 4763 | |||
GenBank mRNA | AB037814.1 | - | 4 | 4605 | ||
BC063551.1 | - | 1 | 788 | |||
BC136606.1 | - | 5 | 4648 | 9020596 | ||
BC136607.1 | - | 5 | 4648 | 9020597 |
Gene Symbol: HMGN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005517.3 | NP_005508.1 | 6 | 723 | ||
GenBank mRNA | BC000378.1 | - | 6 | 626 | 2820349 | |
BC003050.1 | - | 6 | 617 | 3543809 | ||
BC003689.1 | - | 6 | 587 | |||
BC007124.1 | - | 1 | 413 | |||
BC014644.1 | - | 6 | 606 | 3355317 | ||
BC020307.1 | - | 1 | 217 | |||
BC032140.2 | - | 6 | 616 | 5021546 | ||
BC061898.1 | - | 1 | 351 | |||
BC064347.1 | - | 1 | 393 | |||
BC070297.1 | - | 2 | 576 | 4282630 | ||
BC071707.1 | - | 6 | 616 | 3849426 | ||
BC072010.1 | - | 6 | 612 | 6273507 | ||
BC072011.1 | - | 6 | 605 | 6647282 | ||
BC075837.1 | - | 6 | 709 | 6272425 | ||
BC081567.1 | - | 6 | 610 | 4131303 | ||
BC110390.1 | - | 6 | 619 | 5190365 | ||
M12623.1 | - | 6 | 609 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
