siRNA Details
Gene Symbol: ZNF841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136499.1 | NP_001129971.1 | 7 | 1660 | ||
NM_001321349.1 | NP_001308278.1 | 5 | 1404 | |||
XM_011526833.2 | XP_011525135.1 | 6 | 1552 | |||
XM_017026652.1 | XP_016882141.1 | 3 | 1209 | |||
XM_017026653.1 | XP_016882142.1 | 7 | 1618 | |||
XM_017026654.1 | XP_016882143.1 | 7 | 1604 | |||
XM_017026655.1 | XP_016882144.1 | 3 | 1206 | |||
XM_017026656.1 | XP_016882145.1 | 6 | 3255 | |||
XM_017026657.1 | XP_016882146.1 | 6 | 1549 | |||
XM_017026658.1 | XP_016882147.1 | 3 | 1209 | |||
XM_017026659.1 | XP_016882148.1 | 3 | 1209 | |||
XM_017026660.1 | XP_016882149.1 | 3 | 1209 | |||
XM_017026661.1 | XP_016882150.1 | 3 | 1209 | |||
XM_017026662.1 | XP_016882151.1 | 6 | 3255 | |||
XM_017026663.1 | XP_016882152.1 | 6 | 1395 | |||
GenBank mRNA | AK093512.1 | - | 1 | 141 | ||
AK129938.1 | - | 1 | 887 | |||
AK131334.1 | - | 1 | 547 | |||
AK131409.1 | - | 5 | 1404 | |||
BC136254.1 | - | 4 | 1250 | 9020241 |
Gene Symbol: ZNF285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291488.1 | NP_001278417.1 | 5 | 1053 | ||
NM_001291489.1 | NP_001278418.1 | 4 | 1123 | |||
NM_001291490.1 | NP_001278419.1 | 5 | 1187 | |||
NM_001291491.1 | NP_001278420.1 | 3 | 905 | |||
NM_152354.5 | NP_689567.4 | 4 | 1032 | |||
GenBank mRNA | AK055309.1 | - | 1 | 641 | ||
AL833447.1 | - | 4 | 1010 | |||
AY166790.1 | - | 2 | 822 | |||
BC069384.1 | - | 1 | 551 | 7262358 | ||
BC074824.2 | - | 1 | 514 | |||
BC074875.2 | - | 1 | 514 | 30915235 | ||
BC117300.1 | - | 3 | 996 | 40125851 | ||
BC117302.1 | - | 3 | 996 | 40125853 | ||
BC143984.1 | - | 4 | 1017 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001177570, BC083001.
Gene Ontology Categories:
