siRNA Details
Gene Symbol: ZNF43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256648.1 | NP_001243577.1 | 7 | 2468 | ||
NM_001256649.1 | NP_001243578.1 | 7 | 2333 | |||
NM_001256650.1 | NP_001243579.1 | 6 | 2205 | |||
NM_001256651.1 | NP_001243580.1 | 3 | 1819 | |||
NM_001256653.1 | NP_001243582.1 | 4 | 1946 | |||
NM_001256654.1 | NP_001243583.1 | 3 | 1828 | |||
NM_003423.3 | NP_003414.2 | 4 | 1955 | |||
XM_011528257.2 | XP_011526559.1 | 4 | 1877 | |||
XM_011528259.2 | XP_011526561.1 | 4 | 2434 | |||
XM_017027207.1 | XP_016882696.1 | 7 | 7946 | |||
XM_017027208.1 | XP_016882697.1 | 7 | 2626 | |||
XM_017027209.1 | XP_016882698.1 | 7 | 7679 | |||
XM_017027210.1 | XP_016882699.1 | 6 | 7470 | |||
XM_017027211.1 | XP_016882700.1 | 7 | 7613 | |||
XM_017027212.1 | XP_016882701.1 | 6 | 2473 | |||
XM_017027213.1 | XP_016882702.1 | 7 | 7721 | |||
XM_017027214.1 | XP_016882703.1 | 5 | 2396 | |||
XM_017027215.1 | XP_016882704.1 | 6 | 2235 | |||
XM_017027216.1 | XP_016882705.1 | 5 | 2021 | |||
GenBank mRNA | AK291353.1 | - | 7 | 2283 | ||
BC006528.2 | - | 4 | 1922 | 3687640 | ||
BT009756.1 | - | 3 | 1767 | |||
CR933640.1 | - | 7 | 2428 | |||
X59244.1 | - | 6 | 2104 |
Gene Symbol: LOC400682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011528477.2 | XP_011526779.2 | 4 | 1038 | ||
GenBank mRNA | BX647610.1 | - | 2 | 841 |
Gene Symbol: ZNF100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173531.3 | NP_775802.2 | 5 | 1405 | ||
GenBank mRNA | AK126551.1 | - | 5 | 1356 | ||
BC031248.1 | - | 4 | 1370 | |||
BC035579.1 | - | 5 | 1317 | 5576923 | ||
BX640982.1 | - | 1 | 115 |
Gene Symbol: ZNF430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172671.1 | NP_001166142.1 | 5 | 1384 | ||
NM_025189.3 | NP_079465.3 | 5 | 1387 | |||
GenBank mRNA | AK023721.1 | - | 5 | 1366 | ||
AK290475.1 | - | 5 | 1387 | |||
AL832422.1 | - | 1 | 272 | |||
AY269787.1 | - | 5 | 1316 | |||
BC111776.1 | - | 5 | 1272 | 40033720 |
Gene Symbol: ZNF492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020855.2 | NP_065906.1 | 4 | 1174 | ||
GenBank mRNA | AB040906.1 | - | 4 | 1061 |
Gene Symbol: ZNF626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076675.2 | NP_001070143.1 | 4 | 1512 | ||
GenBank mRNA | AK096221.1 | - | 3 | 1238 | ||
BC150651.1 | - | 5 | 1423 | 9057022 | ||
BX648764.1 | - | 3 | 1265 | |||
CR749233.1 | - | 5 | 1464 |
Gene Symbol: ZNF714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182515.3 | NP_872321.2 | 5 | 2299 | ||
NR_117086.1 | 6 | 2318 | ||||
NR_117087.1 | 5 | 2299 | ||||
GenBank mRNA | BC022527.1 | - | 6 | 2260 | ||
BC037782.1 | - | 6 | 2265 | 4797729 | ||
BC050468.2 | - | 5 | 2237 |
Gene Symbol: ZNF728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267716.1 | NP_001254645.1 | 4 | 1957 |
Gene Symbol: ZNF738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027130.1 | 5 | 8993 |
Gene Symbol: ZNF98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098626.1 | NP_001092096.1 | 4 | 1175 | ||
XM_011527705.1 | XP_011526007.1 | 4 | 1134 | |||
GenBank mRNA | BC110576.1 | - | 3 | 965 | 40007052 | |
BC129810.1 | - | 3 | 965 |
Gene Symbol: ZNF99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080409.2 | NP_001073878.2 | 4 | 5662 | ||
GenBank mRNA | BC021822.1 | - | 1 | 474 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
