siRNA Details
Gene Symbol: HLA-DRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019111.4 | NP_061984.2 | 4 | 815 | ||
GenBank mRNA | AK301134.1 | - | 4 | 712 | ||
AK301354.1 | - | 3 | 456 | |||
AK310161.1 | - | 3 | 263 | |||
AK313123.1 | - | 4 | 770 | |||
BC032350.1 | - | 4 | 746 | 5214156 | ||
BC071659.1 | - | 4 | 754 | 5162925 | ||
BG548634.1 | - | 3 | 441 | |||
CR457013.1 | - | 4 | 706 | |||
J00194.1 | - | 4 | 732 | |||
K01171.1 | - | 4 | 720 | |||
M60333.1 | - | 4 | 658 | |||
M60334.1 | - | 4 | 733 | |||
V00523.1 | - | 3 | 615 | |||
V00528.1 | - | 1 | 38 |
Gene Symbol: HLA-DQA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF522250.1 | - | 726 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
