siRNA Details
Gene Symbol: HLA-DRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019111.4 | NP_061984.2 | 2 | 273 | ||
GenBank mRNA | AK297032.1 | - | 2 | 236 | ||
AK301134.1 | - | 2 | 245 | |||
AK301190.1 | - | 2 | 228 | |||
AK301354.1 | - | 2 | 231 | |||
AK310161.1 | - | 2 | 202 | |||
AK313123.1 | - | 2 | 228 | |||
BC032350.1 | - | 2 | 204 | 5214156 | ||
BC071659.1 | - | 2 | 212 | 5162925 | ||
BM849755.1 | - | 2 | 274 | |||
CR457013.1 | - | 2 | 164 | |||
J00194.1 | - | 2 | 190 | |||
K01171.1 | - | 2 | 178 | |||
M60333.1 | - | 2 | 191 | |||
M60334.1 | - | 2 | 191 | |||
V00523.1 | - | 1 | 73 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
