siRNA Details
Gene Symbol: HLA-DRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019111.4 | NP_061984.2 | 5 | 1089 | ||
GenBank mRNA | AK297032.1 | - | 5 | 740 | ||
AK301134.1 | - | 5 | 986 | |||
AK301354.1 | - | 4 | 730 | |||
AK304429.1 | - | 4 | 457 | |||
AK310161.1 | - | 4 | 537 | |||
BC032350.1 | - | 5 | 1020 | 5214156 | ||
BC071659.1 | - | 5 | 1028 | 5162925 | ||
J00194.1 | - | 5 | 1007 | |||
K01171.1 | - | 5 | 994 | |||
V00523.1 | - | 4 | 889 | |||
V00528.1 | - | 2 | 312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
