siRNA Details
Gene Symbol: ZNF706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042510.1 | NP_001035975.1 | 3 | 484 | ||
NM_001267708.1 | NP_001254637.1 | 4 | 528 | |||
NM_001267709.1 | NP_001254638.1 | 2 | 282 | |||
NM_016096.3 | NP_057180.1 | 2 | 369 | |||
GenBank mRNA | AF125099.1 | - | 2 | 205 | ||
AF275802.1 | - | 2 | 250 | |||
AF275809.1 | - | 2 | 250 | |||
AK027802.1 | - | 2 | 1593 | |||
AK290477.1 | - | 2 | 253 | |||
AV721205.1 | - | 3 | 484 | |||
BC015925.1 | - | 1 | 126 | 3909510 | ||
BC068524.1 | - | 2 | 253 | 5263977 | ||
BG704587.1 | - | 2 | 287 | |||
BI669563.1 | - | 4 | 483 | |||
BI755311.1 | - | 3 | 392 | |||
DB231109.1 | - | 2 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
