siRNA Details
Gene Symbol: ZNF706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042510.1 | NP_001035975.1 | 3 | 436 | ||
NM_001267708.1 | NP_001254637.1 | 4 | 480 | |||
NM_001267709.1 | NP_001254638.1 | 2 | 234 | |||
NM_016096.3 | NP_057180.1 | 2 | 321 | |||
GenBank mRNA | AF125099.1 | - | 2 | 157 | ||
AF275802.1 | - | 2 | 202 | |||
AF275809.1 | - | 2 | 202 | |||
AK027802.1 | - | 2 | 1545 | |||
AK290477.1 | - | 2 | 205 | |||
AV721205.1 | - | 3 | 436 | |||
BC015925.1 | - | 1 | 78 | 3909510 | ||
BC068524.1 | - | 2 | 205 | 5263977 | ||
BG704587.1 | - | 2 | 239 | |||
BI669563.1 | - | 4 | 435 | |||
BI755311.1 | - | 3 | 344 | |||
DB231109.1 | - | 2 | 234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
