siRNA Details
Gene Symbol: RPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000971.3 | NP_000962.2 | 1 | 689 | ||
XM_006716463.2 | XP_006716526.1 | 6 | 1282 | |||
GenBank mRNA | AK225246.1 | - | 1 | 688 | ||
AK225254.1 | - | 1 | 687 | |||
AK291119.1 | - | 6 | 687 | |||
AK313365.1 | - | 6 | 687 | |||
BC006095.1 | - | 1 | 677 | 2821364 | ||
BC008850.2 | - | 1 | 714 | 4138113 | ||
BC009599.1 | - | 1 | 689 | 4134557 | ||
BC071671.1 | - | 1 | 670 | 4392472 | ||
BC071894.1 | - | 1 | 674 | 2906310 | ||
BC071895.1 | - | 1 | 672 | 6568654 | ||
BC087837.1 | - | 1 | 673 | 6170371 | ||
CR456773.1 | - | 1 | 667 | |||
L16558.1 | - | 1 | 667 | |||
X52967.1 | - | 6 | 688 | |||
X57958.1 | - | 1 | 704 | |||
X57959.1 | - | 1 | 677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
