siRNA Details
Gene Symbol: XAGE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130776.1 | NP_570132.1 | 3 | 179 | ||
NM_133179.2 | NP_573440.1 | 3 | 210 | |||
XM_011530768.2 | XP_011529070.1 | 3 | 276 | |||
XM_011530769.2 | XP_011529071.1 | 3 | 239 | |||
XM_011530770.2 | XP_011529072.1 | 3 | 203 | |||
GenBank mRNA | AF318372.1 | - | 3 | 191 | ||
AJ318881.1 | - | 3 | 179 | |||
BC062680.1 | - | 3 | 214 | 4619720 |
Gene Symbol: XAGE-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ318895.1 | - | 2 | 70 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
