siRNA Details
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 5 | 397 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 9 | 1355 |
Gene Symbol: PMS2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047753.1 | - | 4 | 290 |
Gene Symbol: PMS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003614.3 | 4 | 314 | |||
GenBank mRNA | DA215155.1 | - | 4 | 301 |
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 5 | 442 | |||
NR_027776.2 | 4 | 339 | ||||
NR_027777.2 | 3 | 252 | ||||
GenBank mRNA | AK309660.1 | - | 4 | 287 | ||
BC052588.1 | - | 3 | 255 | |||
BF984173.1 | - | 4 | 338 | |||
D38436.1 | - | 5 | 423 | |||
D38439.1 | - | 3 | 251 | |||
D38501.1 | - | 3 | 251 | |||
D38502.1 | - | 5 | 423 |
Gene Symbol: PMS2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017005.1 | - | 6 | 604 |
Gene Symbol: PMS2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130940.1 | 5 | 468 | |||
GenBank mRNA | AB017006.1 | - | 5 | 563 | ||
DB472687.1 | - | 5 | 468 |
Gene Symbol: PMS2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028058.1 | 3 | 730 | |||
GenBank mRNA | BM845848.1 | - | 4 | 271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
