siRNA Details
Gene Symbol: ANKRD20A8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003366.2 | 16, 17 | 2631 |
Gene Symbol: ANKRD18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244752.1 | NP_001231681.1 | 12, 13 | 2348 | ||
XM_011517881.2 | XP_011516183.1 | 13, 14 | 2415 | |||
XM_011517883.2 | XP_011516185.1 | 12, 13 | 2358 | |||
XM_011517885.2 | XP_011516187.1 | 7, 8 | 1503 | |||
XM_017014728.1 | XP_016870217.1 | 7, 8 | 1500 | |||
GenBank mRNA | DW421358.1 | - | 1, 2 | 47 |
Gene Symbol: ANKRD18CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136286.1 | 14, 15 | 2512 |
Gene Symbol: LOC644249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015390.1 | XP_016870879.1 | 2, 3 | 206 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
