siRNA Details
Gene Symbol: KRTAP1-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030967.2 | NP_112229.1 | 1 | 87 | ||
GenBank mRNA | AB052868.1 | - | 2 | 87 | ||
AB052934.1 | - | 1 | 87 | |||
AB055057.1 | - | 1 | 38 | |||
AJ406926.1 | - | 1 | 56 | |||
BC069321.1 | - | 1 | 73 | |||
BC069450.1 | - | 1 | 73 | |||
BC105069.1 | - | 1 | 62 | 8144072 | ||
BC105071.1 | - | 1 | 62 | 8144074 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
