siRNA Details
Gene Symbol: KRTAP1-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031957.1 | NP_114163.1 | 1 | 396 | ||
GenBank mRNA | AJ406928.1 | - | 1 | 396 | ||
BC069533.1 | - | 1 | 396 | 7262171 | ||
BC093841.1 | - | 1 | 422 | 7939686 | ||
BC101555.1 | - | 1 | 422 | 8069061 |
Gene Symbol: KRTAP1-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030967.2 | NP_112229.1 | 1 | 413 | ||
GenBank mRNA | AB052868.1 | - | 2 | 275 | ||
AB052934.1 | - | 1 | 413 | |||
AB055057.1 | - | 2 | 88 | |||
AJ406926.1 | - | 1 | 382 | |||
BC069321.1 | - | 1 | 399 | |||
BC069450.1 | - | 1 | 399 | |||
BC105069.1 | - | 1 | 388 | 8144072 | ||
BC105071.1 | - | 1 | 388 | 8144074 |
Gene Symbol: KRTAP1-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030966.1 | NP_112228.1 | 1 | 353 | ||
GenBank mRNA | AJ406927.1 | - | 1 | 353 | ||
BC069161.1 | - | 1 | 352 | 7216931 | ||
BC101150.1 | - | 1 | 347 | 40015405 | ||
BC101151.2 | - | 1 | 347 | 40015406 | ||
BC101152.1 | - | 1 | 377 | 40015409 | ||
BC101153.2 | - | 1 | 347 | 40015410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
